4-Bromophenylhydrazinium chloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS21279G25
ACTIAS21279G25EA 0 GBP
ACTIAS21279G25
4-Bromophenylhydrazinium chloride
4-Bromophenylhydrazinium chloride
Formula: C₆H₈BrClN₂
MDL Number: MFCD00012941
CAS Number: 622-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR