Imidazo[1,2-a]pyridine-3-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS21164G1 AS21164G5
ACTIAS21164G1EA 0 GBP
ACTIAS21164G1 ACTIAS21164G5
Imidazo[1,2-a]pyridine-3-carboxylic acid
Imidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₈H₆N₂O₂
MDL Number: MFCD01720458
CAS Number: 6200-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR