3-Sulphamoylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS20647G5
ACTIAS20647G5EA 0 GBP
ACTIAS20647G5
3-Sulphamoylbenzoic acid
3-Sulphamoylbenzoic acid
Formula: C₇H₇NO₄S
MDL Number: MFCD01122318
CAS Number: 636-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR