3-Morpholinobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS20585G1 AS20585G5
ACTIAS20585G1EA 0 GBP
ACTIAS20585G1 ACTIAS20585G5
3-Morpholinobenzaldehyde
3-Morpholinobenzaldehyde
Formula: C₁₁H₁₃NO₂
MW: 191.2264 g/mol
MDL Number: MFCD07772811
CAS Number: 446866-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR