4-Chloro-6,7-dimethoxyquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS20461G25 AS20461G5
ACTIAS20461G25EA 0 GBP
ACTIAS20461G25 ACTIAS20461G5
4-Chloro-6,7-dimethoxyquinazoline
4-Chloro-6,7-dimethoxyquinazoline
Formula: C₁₀H₉ClN₂O₂
MW: 224.65 g/mol
MDL Number: MFCD01570172
CAS Number: 13790-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR