p-Toluonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Methylbenzonitrile, p-Methylbenzonitrile

AS20269G100
ACTIAS20269G100EA 82 GBP
ACTIAS20269G100
p-Toluonitrile
p-Toluonitrile
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 217.6 °C (1013 hPa)
Melting Pt: 25…28 °C
Density: 0.98 g/cm³ (20 °C)
MDL Number: MFCD00001827
CAS Number: 104-85-8
EINECS: 203-244-8
Merck Index: 12,09676

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR