(2,4,5-Trifluorophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AS20228G100 AS20228G25
ACTIAS20228G100EA 0 GBP
ACTIAS20228G100 ACTIAS20228G25
(2,4,5-Trifluorophenyl)acetic acid
(2,4,5-Trifluorophenyl)acetic acid
Formula: (C₆H₂F₃)CH₂COOH
MDL Number: MFCD00082479
CAS Number: 209995-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR