(±)-β-Aminophenethyl alcohol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol

AS20221G100 AS20221G25
ACTIAS20221G100EA 0 GBP
ACTIAS20221G100 ACTIAS20221G25
(±)-β-Aminophenethyl alcohol
(±)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00130145
CAS Number: 7568-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR