Sodium chloride

Supplier: ACTIVE MOTIF
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Saline, NaCl, Sea salt, Halite

Z15081092.1L
ACSDZ15081092.1LEA 103 GBP
ACSDZ15081092.1L
Sodium chloride
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 100 °C
Melting Pt: 801
Density: 1 g/cm³
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR