Potassium bromide 99,99%, HighPurS-CERT

Supplier: ACTIVE MOTIF
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

PSPBRC.100G
ACSDPSPBRC.100GEA 617 GBP
ACSDPSPBRC.100G
Potassium bromide 99,99%, HighPurS-CERT
Potassium bromide

Certifications: ISO 17034, ISO 17025, ISO 9001.

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C
Melting Pt: 734 °C
Density: 1,4 g/cm³
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR