Nitrite-Nitrogen standard solution, 1000 mg/L NO₂- (as N) in Water standard for ion chromatography
Supplier: ACTIVE MOTIF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H043.W.L1 H043.W.L25 H043.W.L5
ACSDH043.W.L1EA 0 GBP
ACSDH043.W.L1 ACSDH043.W.L25 ACSDH043.W.L5
Nitrite-Nitrogen standard solution, 1000 mg/L NO₂- (as N) in Water standard for ion chromatography
Standards Anionic Standards Nitrite Standards