Lithium standard solution, 1000 mg/L Li in water standard for ion chromatography
Supplier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H008.W.L1 H008.W.L25 H008.W.L5
ACSDH008.W.L1EA 114 GBP
ACSDH008.W.L1 ACSDH008.W.L25 ACSDH008.W.L5
Lithium standard solution, 1000 mg/L Li in water standard for ion chromatography
Standards Cationic Standards Lithium Standards