Lutetium standard solution, 10000 mg/L Lu in 5% nitric acid standard for ICP
Supplier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C131.5NP.L1 C131.5NP.L25 C131.5NP.L5
ACSDC131.5NP.L1EA 1940 GBP
ACSDC131.5NP.L1 ACSDC131.5NP.L25 ACSDC131.5NP.L5
Lutetium standard solution, 10000 mg/L Lu in 5% nitric acid standard for ICP
Standards Cationic Standards Lutetium Standards