Iron standard solution, 1000 mg/L Fe in dil. nitric acid standard for ICP
Supplier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C019.2NP.L1 C019.2NP.L25 C019.2NP.L5
ACSDC019.2NP.L1EA 110 GBP
ACSDC019.2NP.L1 ACSDC019.2NP.L25 ACSDC019.2NP.L5
Iron standard solution, 1000 mg/L Fe in dil. nitric acid standard for ICP
Standards Cationic Standards Iron Standards