Cesium standard solution, 1,000 mg/L Cs in water standard for ICP
Supplier: ACTIVE MOTIF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C014.W.L1 C014.W.L25 C014.W.L5
ACSDC014.W.L1EA 110 GBP
ACSDC014.W.L1 ACSDC014.W.L25 ACSDC014.W.L5
Cesium standard solution, 1,000 mg/L Cs in water standard for ICP
Standards Cationic Standards Cesium Standards