Aluminium standard solution, 1000 mg/L Al in 2% hydrochloric acid standard for ICP
Supplier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C002.2CP.L1 C002.2CP.L25 C002.2CP.L5
ACSDC002.2CP.L1EA 110 GBP
ACSDC002.2CP.L1 ACSDC002.2CP.L25 ACSDC002.2CP.L5
Aluminium standard solution, 1000 mg/L Al in 2% hydrochloric acid standard for ICP
Standards Cationic Standards Aluminium Standards