Duloxetine hydrochloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (S)-(+)-N-Methyl-3-(1-naphthyloxy)-3-(2-thienyl)propylamine Hydrochloride, (S)-Duloxetine hydrochloride

463140010 463140050
ACRO463140010EA 51 GBP
ACRO463140010 ACRO463140050
Duloxetine hydrochloride
Duloxetine hydrochloride

  • Bioactive

Formula: C₁₈H₁₉NOS
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD06407958
CAS Number: 136434-34-9

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 162 to 168 °C
Loss on drying ≤0.5 % (105 °C, 3 h)
HPLC ≥97.5 %
Specific optical rotation +116° to +125° (20 °C, 589 nm) (1 % in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR