6-tert-Butyl-2,4-xylenol 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-(tert-Butyl)-4,6-dimethylphenol

462570100 462570500
ACRO462570100EA 30.2 GBP
ACRO462570100 ACRO462570500
6-tert-Butyl-2,4-xylenol 97%
6-tert-Butyl-2,4-xylenol
Formula: C₁₂H₁₈O
MW: 178.28 g/mol
Boiling Pt: 249 °C
Melting Pt: 22…23 °C
Storage Temperature: Ambient
MDL Number: MFCD00002234
CAS Number: 1879-09-0
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Appearance White to light yellow Liquid or solid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR