Mycophenolic acid 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

459380050 459380250 459380010
ACRO459380050EA 68.5 GBP
ACRO459380050 ACRO459380250 ACRO459380010
Mycophenolic acid 98%
Mycophenolic acid
Formula: C₁₇H₂₀O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS Number: 24280-93-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 139 to 145 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR