Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

458230010 458230050 458230250
ACRO458230010EA 90.4 GBP
ACRO458230010 ACRO458230050 ACRO458230250
Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed
Polyvinyl alcohol
Formula: (CH₂CHOH)n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Viscosity 28 to 32 cP (4 % in water)
Degree of hydrolysis 99.3 to 100.0 mol %
Ash ≤1.2 % (as Na2O)
pH 5.5 to 7.5 (4 % in water)
Volatile matter ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR