Iodine, meetSpec_BP,Ph.Eur.,USP

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

457760010 457760500 457762500
ACRO457760010EA 147 GBP
ACRO457760010 ACRO457760500 ACRO457762500
Iodine, meetSpec_BP,Ph.Eur.,USP
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 185 °C (760 mmHg)
Melting Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now

Specification Test Results

Appearance Greyish-violet Brittle plates or crystals with metallic sheen
Titration Iodometric 99.8 to 100.5 %
Non-volatile matter ≤0.05 %
Chloride and Bromide ≤250 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR