3,4-Ethylenedioxythiophene 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

454930050 454930250
ACRO454930050EA 48.1 GBP
ACRO454930050 ACRO454930250
3,4-Ethylenedioxythiophene 99%
3,4-Ethylenedioxythiophene
Formula: C₆H₆O₂S
Boiling Pt: 193 °C
Melting Pt: 10.5…12 °C
Density: 1.331 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD02093622
CAS Number: 126213-50-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR