2-Hydroxy-6-methylbenzoic acid 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2,6-Cresotic acid, 6-Hydroxy-o-toluic acid, 6-Methylsalicylic acid

453060010 453060050 453060250
ACRO453060010EA 39.7 GBP
ACRO453060010 ACRO453060050 ACRO453060250
2-Hydroxy-6-methylbenzoic acid 98%
2-Hydroxy-6-methylbenzoic acid
Formula: C₈H₈O₃
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD01194284
CAS Number: 567-61-3

Order Now

Specification Test Results

Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
Melting point 168 to 176 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR