Quartz, fine wool 4 µm

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

451040100 451040500
ACRO451040100EA 34.6 GBP
ACRO451040100 ACRO451040500
Quartz, fine wool 4 µm
Quartz
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00217788
CAS Number: 14808-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5273273c3b2e473c6ced4e97e18a338f3d21302c&vid=9c414ebb18d830bfa8a77b318dbbb0e6a96da07b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Fibers


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR