Hydrogen chloride 4->6 N in 2-propanol, technical

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

450720010 450720025
ACRO450720010EA 33 GBP
ACRO450720010 ACRO450720025
Hydrogen chloride 4->6 N in 2-propanol, technical
Hydrogen chloride
Formula: HCl
Density: 0.909 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Water ≤5 %
Strength 4 to 6 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR