Ciprofloxacin 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

449620050 449620250
ACRO449620050EA 28.2 GBP
ACRO449620050 ACRO449620250
Ciprofloxacin 98%
Ciprofloxacin
Formula: C₁₇H₁₈FN₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00185755
CAS Number: 85721-33-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Sulfated ash ≤0.1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR