Di-tert-butyl phosphonate 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

449390010 449390050 449390250
ACRO449390010EA 16 GBP
ACRO449390010 ACRO449390050 ACRO449390250
Di-tert-butyl phosphonate 96%
Di-tert-butyl phosphonate
Formula: C₈H₁₉O₃P
Boiling Pt: 66…68 °C (.5 mmHg)
Flash Pt: 163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00995019
CAS Number: 13086-84-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20°C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR