Mercury(II) bromide ACS

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Mercuric bromide, Mercury dibromide

448121000
ACRO448121000EA 62.6 GBP
ACRO448121000
Mercury(II) bromide ACS
Mercury (II) bromide
Formula: Br₂Hg
MW: 360.4 g/mol
Boiling Pt: 322 °C (760 mmHg)
Melting Pt: 236 °C
Density: 6,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011040
CAS Number: 7789-47-1
UN: 1634
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to very light yellow Powder
Titration Complexometric ≥98.0 %
Chloride (Cl) ≤0.25 %
Insoluble matter ≤0.05 % (In methanol)
Residue after reduction ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR