Potassium thiocyanate 99% for biochemistry

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Potassium rhodanide

448000010 448002500
ACRO448000010EA 113 GBP
ACRO448000010 ACRO448002500
Potassium thiocyanate 99% for biochemistry
Potassium thiocyanate
Formula: KSCN
MW: 97.18 g/mol
Boiling Pt: 500
Melting Pt: 170…179 °C
Density: 1.886 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
Merck Index: 12,07866

Order Now

Specification Test Results

Appearance White Crystals
Titration Argentometric ≥98.5 %
pH 5.3 to 8.7
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.0005 %
Lead (Pb) ≤0.001 %
Sodium (Na) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR