Sodium sulphite 98% Biochemistry

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

447950010 447952500
ACRO447950010EA 97.5 GBP
ACRO447950010 ACRO447952500
Sodium sulphite 98% Biochemistry
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.63 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
Merck Index: 13,08757

Order Now

Specification Test Results

Appearance White Powder or crystals
Titration Iodimetric ≥97.5 %
Solubility (1 M in water) Clear colorless
Chloride (Cl) ≤0.02 %
Arsenic (As) ≤1 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Selenium (Se) ≤10 ppm
Zinc (Zn) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR