1-Propanol ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

445770010 445775000
ACRO445770010EA 41.8 GBP
ACRO445770010 ACRO445775000
1-Propanol ACS
1-Propanol
Formula: H₃CCH₂CH₂OH
Boiling Pt: 97 °C (760 mmHg)
Melting Pt: –127 °C
Density: 0.8 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
UN: 1274
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.2 %
Solubility (in water) Passes test
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.03 % (as C2H5CHO)
Isopropyl alcohol ≤0.05 %
Titrable acid ≤0.0004 meq/g
Ethanol ≤0.01 %
Methanol ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR