5-(Methylsulphonyl)pyridine-3-boronic acid pinacol ester 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

445180010
ACRO445180010EA 217 GBP
ACRO445180010
5-(Methylsulphonyl)pyridine-3-boronic acid pinacol ester 95%
5-(Methylsulphonyl)pyridine-3-boronic acid pinacol ester
Formula: C₁₂H₁₈BNO₄S
Melting Pt: 164…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12198145
CAS Number: 1206641-26-0

Order Now

Specification Test Results

Appearance Off-white to tan Solid
Melting point 164 to 169°C
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR