Agar, Bacteriological Grade

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Agar Agar

443570010 443570050 443572500
ACRO443570010EA 214 GBP
ACRO443570010 ACRO443570050 ACRO443572500
Agar, Bacteriological Grade
Agar
Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to cream to light yellow to beige Powder
Infrared spectrum Conforms
Loss on drying ≤15 % (1 g, 105°C)
Solubility (1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution
Gel strength ≥700 g/cm² (1.5 %)
Gel point 32 to 38°C (1.5 %)
Ash ≤5 %
pH 6.0 to 7.5 (10 % suspension)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR