2-Chloro-1,3-dimethylimidazolinium chloride 90%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: DMC

439520050 439520250 439521000
ACRO439520050EA 48.1 GBP
ACRO439520050 ACRO439520250 ACRO439521000
2-Chloro-1,3-dimethylimidazolinium chloride 90%
2-Chloro-1,3-dimethylimidazolinium chloride
Formula: C₅H₁₀Cl₂N₂
MW: 169.05 g/mol
Melting Pt: 112…121 °C
Storage Temperature: Ambient
MDL Number: MFCD09039290
CAS Number: 37091-73-9

Order Now

Specification Test Results

Appearance White to off-white to brown Crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥88.0 %
Elemental Analysis 14.58 to 18.56 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR