Calcium carbonate 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Marble

437190050 437190250
ACRO437190050EA 37 GBP
ACRO437190050 ACRO437190250
Calcium carbonate 99.999% (trace metals basis)
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C (dec.)
Density: 2,71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric 39 to 42 %
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR