Sodium nitrate 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

437160100 437160500 437165000
ACRO437160100EA 51.2 GBP
ACRO437160100 ACRO437160500 ACRO437165000
Sodium nitrate 99.999% (trace metals basis)
Sodium nitrate
Formula: NaNO₃
Boiling Pt: 380 °C (760 mmHg)
Melting Pt: 306 °C
Density: 2,26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
UN: 1498
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White Powder or crystals
Total trace metal impurities ≤10 ppm
Sodium (Na) Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR