Selenious acid 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Selenous acid

437120500
ACRO437120500EA 107 GBP
ACRO437120500
Selenious acid 99.999% (trace metals basis)
Selenious acid
Formula: H₂O₃Se
MW: 128.97 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 70 °C (dec.)
Density: 1.998
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
UN: 3283
ADR: 6.1 II
Merck Index: 14,08430

Order Now

Specification Test Results

Appearance White Crystals
Assay 60 to 62.4 % (Se)
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR