Sodium thiosulphate pentahydrate 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

436940250 436941000
ACRO436940250EA 126 GBP
ACRO436940250 ACRO436941000
Sodium thiosulphate pentahydrate 99.999% (trace metals basis)
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
Melting Pt: 48.5 °C
Density: 1.72 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodimetric ≥98.5 %
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR