Ammonium dihydrogen phosphate 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

436790250 436791000 436795000
ACRO436790250EA 44.7 GBP
ACRO436790250 ACRO436791000 ACRO436795000
Ammonium dihydrogen phosphate 99.999% (trace metals basis)
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
Melting Pt: 190 °C
Density: 1,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1

Order Now

Specification Test Results

Appearance Colorless to white Crystals
Total trace metal impurities ≤10 ppm
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR