1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

435202500
ACRO435202500EA 218 GBP
ACRO435202500
1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
1,2-Bis((2R,5R)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Formula: C₃₄H₅₄BF₄P₂Rh
Storage Temperature: Ambient
MDL Number: MFCD07369039
CAS Number: 569650-64-2

Order Now

Specification Test Results

Appearance Yellow to orange to red to brown Powder
Infrared spectrum Conforms
NMR (1H-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 54.7 to 59.3 % C
Specific optical rotation -78° to -86° (20°C, 589 nm) (c=0.8 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR