Ytterbium(III) trifluoromethanesulphonate, anhydrous 99.9% (trace metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ytterbium (III) triflate

434050010 434050050
ACRO434050010EA 26.6 GBP
ACRO434050010 ACRO434050050
Ytterbium(III) trifluoromethanesulphonate, anhydrous 99.9% (trace metals basis)
Ytterbium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Yb
Storage Temperature: Ambient
MDL Number: MFCD06200261
CAS Number: 54761-04-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Complexometric 26.8 to 29.0 % (Yb)
Total conc. of imp.as REO ≤1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR