4,4,4-Trifluorobutylamine 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-Butanamine,4,4,4-trifluoro-, 4,4,4-trifluorobutan-1-amine, 1-Amino-4,4,4-trifluorobutane

432590010 432590050
ACRO432590010EA 79.2 GBP
ACRO432590010 ACRO432590050
4,4,4-Trifluorobutylamine 97%
4,4,4-Trifluorobutylamine
Formula: C₄H₈F₃N
Boiling Pt: 90…94 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD06213030
CAS Number: 819-46-5
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Titration RCOCl ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR