(R)-(-)-Linalool 95% enantiomers_sum

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool

428311000 428310250
ACRO428311000EA 60.6 GBP
ACRO428311000 ACRO428310250
(R)-(-)-Linalool 95% enantiomers_sum
(R)-(-)-Linalool
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 196…198 °C (760 mmHg)
Density: 0.862 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00135469
CAS Number: 126-91-0
EINECS: 204-811-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4640 (20°C, 589 nm)
GC ≥94 %
Specific gravity 0.859 to 0.864
Specific optical rotation -15° to -21° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR