1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 90%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: tert-Butyl 4-(aminomethyl)benzylcarbamate

426900010 426900050
ACRO426900010EA 50.7 GBP
ACRO426900010 ACRO426900050
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene 90%
1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene

Appearance: White to brown Crystalline powder or solid

Formula: C₁₃H₂₀N₂O₂
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD02683058
CAS Number: 108468-00-4
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or solid
Infrared spectrum Conforms
GC ≥89.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR