Dimethyl chlorophosphate 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

426140050 426140250
ACRO426140050EA 43 GBP
ACRO426140050 ACRO426140250
Dimethyl chlorophosphate 96%
Dimethyl chlorophosphate
Formula: C₂H₆ClO₃P
Boiling Pt: 80 °C (25 mmHg)
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00117904
CAS Number: 813-77-4
UN: 2927
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20°C, 589 nm)
Titration Argentometric ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR