Sodium nitrate 99+% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

424340030 424345000 424340010 424340100
ACRO424340030EA 162 GBP
ACRO424340030 ACRO424345000 ACRO424340010 ACRO424340100
Sodium nitrate 99+% ACS
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C (760 mmHg)
Melting Pt: 306 °C
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1 III
REACH: 01-2119488221-41
Merck Index: 13,08720

Order Now

Specification Test Results

Appearance White Crystals
pH 5.5 to 8.3 (5 % solution at 25.0°C)
Titration after Ion exchange ≥99.0 %
Chloride (Cl) ≤10 ppm
Iodate (IO3) ≤5 ppm
Nitrite (NO2) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤30 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤20 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR