Potassium permanganate 99+% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Permanganic acid potassium salt

424170025 424175000
ACRO424170025EA 170 GBP
ACRO424170025 ACRO424175000
Potassium permanganate 99+% ACS
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Appearance Dark green-black Glistening crystalline powder
Titration with KMnO4 ≥99.0 %
Chloride and Chlorate (as Cl) ≤0.005 %
Sulfate (SO4) ≤0.02 %
Insoluble matter ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR