Potassium nitrate 99+% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Nitrate of Potash

424155000 424150025 424151000
ACRO424155000EA 38 GBP
ACRO424155000 ACRO424150025 ACRO424151000
Potassium nitrate 99+% ACS
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2,109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
pH 4.5 to 8.5 (5% soln. at 25°C)
Titration after Ion exchange ≥99.0 %
Chloride (Cl) ≤0.002 %
Iodate (IO3) ≤5 ppm
Nitrite (NO2) ≤0.001 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.003 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR