Molybdic acid ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

423981000 423985000
ACRO423981000EA 33.9 GBP
ACRO423981000 ACRO423985000
Molybdic acid ACS
Molybdic acid
Formula: H₂MoO₄
MW: 161.95 g/mol
Boiling Pt: 300
Melting Pt: 300
Density: 3.1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration ≥85.0 % (Precipitation titraton of molybdate)
Chloride (Cl) ≤0.002 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.2 %
AsO4, PO4, SiO4 (as SiO2) ≤0.001 %
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.01 % (in dilute NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR