Ammonium iron(III) sulphate dodecahydrate 99% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

423690250 423695000 423690025
ACRO423690250EA 23.3 GBP
ACRO423690250 ACRO423695000 ACRO423690025
Ammonium iron(III) sulphate dodecahydrate 99% ACS
Ammonium iron (III) sulphate dodecahydrate
Formula: NH₄Fe(SO₄)₂·12H₂O
Boiling Pt: 230 °C (760 mmHg)
Melting Pt: 39…41 °C
Density: 1.71 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00150004
CAS Number: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549

Order Now

Specification Test Results

Appearance Violet Crystals
Titration Iodometric 98.5 to 102.0 %
Chloride (Cl) ≤0.001 %
Nitrate (NO3) ≤0.01 %
Calcium (Ca) ≤0.01 %
Copper (Cu) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.003 %
Ferrous iron (Fe 2+) (Limit about 0.001%) Passes test
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR